ContactUsLinkComponent
Precise Viscosity Bath, type WVB-30, capacity 30 Liter, with transparent windows, with digital fuzzy control, temp.range: ambient +5C to 100C, timer: 99 hr. / 59 min., effective space: 330 x 300 x 260 mm, external dimension: 585 x 375 x 420 mm, hea 1 * 1 items
Catalog # WITGDH.WVB01030
Supplier:  witeg Labortechnik
Precise Viscosity Bath, type WVB-30, capacity 30 Liter, with transparent windows, with digital fuzzy control, temp.range: ambient +5C to 100C, timer: 99 hr. / 59 min., effective space: 330 x 300 x 260 mm, external dimension: 585 x 375 x 420 mm, hea 1 * 1 items
Catalog # WITGDH.WVB01030
Supplier:  witeg Labortechnik
Frequently Bought Together