ContactUsLinkComponent
Tube, Specimen Collection and Transport Tube Labcon SuperClear® 5 mL Sterile With attached cap and integral molded elastomeric seal Includes molded graduations and writing surface ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) Validated 1 * 1 items
Catalog # 3813-165-008-9
Supplier:  Labcon
Tube, Specimen Collection and Transport Tube Labcon SuperClear® 5 mL Sterile With attached cap and integral molded elastomeric seal Includes molded graduations and writing surface ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) Validated 1 * 1 items
Catalog # 3813-165-008-9
Supplier:  Labcon
Frequently Bought Together