ContactUsLinkComponent
MiniOne electrophoresis system with carrying case. Includes in the case MiniOne carriage, MiniOne photo hood, MiniOne power supply, MiniOne gel tank, MiniOne casting system and p20 micropipette. 1 * 1 items
Catalog # EMBIM1002-EU
Supplier:  EMBITEC
MiniOne electrophoresis system with carrying case. Includes in the case MiniOne carriage, MiniOne photo hood, MiniOne power supply, MiniOne gel tank, MiniOne casting system and p20 micropipette. 1 * 1 items
Catalog # EMBIM1002-EU
Supplier:  EMBITEC
Frequently Bought Together