ContactUsLinkComponent
MobileALERT for up to 4 alarm notifications notification by SMS. temperature and CO2 alarm (standard), additionally humidity alarm (when equipped with option K7) and O2 alarm (when equipped with option T6) 1 * 1 items
Catalog # MEMMICO150C4
Supplier:  MEMMERT
MobileALERT for up to 4 alarm notifications notification by SMS. temperature and CO2 alarm (standard), additionally humidity alarm (when equipped with option K7) and O2 alarm (when equipped with option T6) 1 * 1 items
Catalog # MEMMICO150C4
Supplier:  MEMMERT
Frequently Bought Together