FDA conforming software AtmoCONTROL (FDA edition). Meets the requirements for the use of electronically stored data sets and electro nic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence f 1 * 1 items
Catalog # MEMMFDAQ1ICO105
Supplier: MEMMERT
Frequently Bought Together