[EN]RACK TEST TUBE HOLES 2-ROW RACKS 2 1 * 1 KS
Catalog # MEMMB48831WTB15
Supplier: MEMMERT
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Rukavice, nitrilové
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Ubrousky čisticí
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals