ContactUsLinkComponent
Embedding molds made out of thin-wall stainless steel for quick temperature adjustment. Pre-warming of molds not necessary. Especial ly designed for use with the Leica embedding cassette program - may, however, be used as well with all common cassett 1 * 12 items
Catalog # LEIH14038832449
Supplier:  LEICA HISTOLOGY
Embedding molds made out of thin-wall stainless steel for quick temperature adjustment. Pre-warming of molds not necessary. Especial ly designed for use with the Leica embedding cassette program - may, however, be used as well with all common cassett 1 * 12 items
Catalog # LEIH14038832449
Supplier:  LEICA HISTOLOGY
Frequently Bought Together