PAPANICOLAOU'S SOLUTION OG 6 SURGIPATH 1 * 5 L
Catalog # LEIH3801660BBE
Supplier: LEICA HISTOLOGY
Frequently Bought Together
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, TECHNICAL
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Kjeldahl tablets, Kjeltabs
Supplier: THOMPSON & CAPPER
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach