ContactUsLinkComponent
Tube, Conical centrifuge Labcon PerformR 15 mL Sterile With attached flat screw caps, molded graduations, and writing surface ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) Validated Sterilization (ISO 11137) for use in Research and Medic 1 * 500 items
Catalog # 3140-335-028-9
Supplier:  Labcon
Tube, Conical centrifuge Labcon PerformR 15 mL Sterile With attached flat screw caps, molded graduations, and writing surface ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) Validated Sterilization (ISO 11137) for use in Research and Medic 1 * 500 items
Catalog # 3140-335-028-9
Supplier:  Labcon
Frequently Bought Together