ContactUsLinkComponent
Tube 0,25ml MiniCollect Complete FX Sodium Fluoride/Potassium Oxalate grey cap pre-assembled with carrier tube 13x75mm sterile 1 * 2.000 items
Catalog # GREI450552_2000
Supplier:  Greiner Bio-One
Tube 0,25ml MiniCollect Complete FX Sodium Fluoride/Potassium Oxalate grey cap pre-assembled with carrier tube 13x75mm sterile 1 * 2.000 items
Catalog # GREI450552_2000
Supplier:  Greiner Bio-One
Frequently Bought Together