ContactUsLinkComponent
Mineral oil standard mixture - 2 components 8g/l each of Mineral Oil [CAS:N/A] Diesel Oil [CAS:N/A] in n-Heptane. Duree de conservation : 24 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 10 mL
Catalog # ACSDF675880
Supplier:  CPACHEM
Mineral oil standard mixture - 2 components 8g/l each of Mineral Oil [CAS:N/A] Diesel Oil [CAS:N/A] in n-Heptane. Duree de conservation : 24 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 10 mL
Catalog # ACSDF675880
Supplier:  CPACHEM
Frequently Bought Together