ContactUsLinkComponent
Homogenizer HG-15A, 27000 rpm, analog internal controller 1 * 1 items
Catalog # WITGDH.WHG02015
Supplier:  witeg Labortechnik
Homogenizer HG-15A, 27000 rpm, analog internal controller 1 * 1 items
Catalog # WITGDH.WHG02015
Supplier:  witeg Labortechnik
Frequently Bought Together