Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]TBN TITRATION SOLVENT 0.4M TEABR 1 * 1 L
Catalog # REGETBNSOLV2F
Supplier:
REAGECON
Certificates
Expand image
[EN]TBN TITRATION SOLVENT 0.4M TEABR 1 * 1 L
Catalog # REGETBNSOLV2F
Supplier:
REAGECON
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Roztoky standardní pro měření vodivosti
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Hydrogenftalát draselný, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Dusičnan 1.000 mg/l ve vodě (z NaNO<sub>3</sub>) Certipur®, Supelco®
Supplier: Merck
Toluen, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Roztoky pufrů v sáčcích, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals