ContactUsLinkComponent
Heliflex AT-1701+ GC Column 25m X 0.25mm X 0.20um 1 * 1 items
Catalog # HISMM8635598
Supplier:  HICHROM
Heliflex AT-1701+ GC Column 25m X 0.25mm X 0.20um 1 * 1 items
Catalog # HISMM8635598
Supplier:  HICHROM
Frequently Bought Together