Ghost Dye Blue 516 is an amine reactive viability dye that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications. This dye irreversibly binds free amines available on the cell surface as well as intracellu 1 * 500 Tests
Catalog # 13-0867-T500
Supplier: Tonbo Biosciences
Frequently Bought Together