ContactUsLinkComponent
Tip Eppendorf-fit Graduated 1000-10000µl Racked Sterile 161mm in length 2 racks of 25 for accurate measurement of large volume solut ions 1 * 50 items
Catalog # ALPCLW7835
Supplier:  Alpha Laboratories
Tip Eppendorf-fit Graduated 1000-10000µl Racked Sterile 161mm in length 2 racks of 25 for accurate measurement of large volume solut ions 1 * 50 items
Catalog # ALPCLW7835
Supplier:  Alpha Laboratories
Frequently Bought Together