Electroporation system ECM 830 bipolar pulse switcher square wave mammalian. Contains generator, pulse switcher, safety dome, cuvett es and cuvette rack. Incorporates automatic bipolar switching into transfection protocols. 1 * 1 items
Catalog # BTXI45-0669
Supplier: BTX
Frequently Bought Together