ContactUsLinkComponent
Pulse switcher. Compatible with ECM 830 and Gemini X2 instruments that have 7 inch touch screens. Allows for automatic execution of gouped pulse bipolar switching protocols when combined with an electroporation waveform generator. 1 * 1 items
Catalog # BTXI45-2061
Supplier:  BTX
undefined
Pulse switcher. Compatible with ECM 830 and Gemini X2 instruments that have 7 inch touch screens. Allows for automatic execution of gouped pulse bipolar switching protocols when combined with an electroporation waveform generator. 1 * 1 items
Catalog # BTXI45-2061
Supplier:  BTX
Frequently Bought Together