ContactUsLinkComponent
TEMPERATURE DATA LOGGER TESTO 174 T STARTER KIT including 3x testo 174T mini data loggers, USB interface for programming and reading the logger, wall bracket, batteries (6 x CR2032 Lithium), CD-ROM with ComSoft Basic software and calibration protoco 1 * 1 KIT
Catalog # 620-2769
Supplier:  Testo
undefined
TEMPERATURE DATA LOGGER TESTO 174 T STARTER KIT including 3x testo 174T mini data loggers, USB interface for programming and reading the logger, wall bracket, batteries (6 x CR2032 Lithium), CD-ROM with ComSoft Basic software and calibration protoco 1 * 1 KIT
Catalog # 620-2769
Supplier:  Testo
Frequently Bought Together