ContactUsLinkComponent
Roller App Bttm Drive 9 Deck Process 230V w/BB ~Speed range 0.3-9.0 RPM ~Data Logging with RS485 Controller ~Remote Access and control ~Battery Back up included 1 * 1 items
Catalog # BELC7320-09230
Supplier:  BELLCO GLASS
Roller App Bttm Drive 9 Deck Process 230V w/BB ~Speed range 0.3-9.0 RPM ~Data Logging with RS485 Controller ~Remote Access and control ~Battery Back up included 1 * 1 items
Catalog # BELC7320-09230
Supplier:  BELLCO GLASS
Frequently Bought Together