ContactUsLinkComponent
Target Protein,,P2RY6 Clonality Polyclonal Application,,WB,ELISA Recommended Dilution,,WB 1:500-2000 ELISA 1:5000-20000 Concentration,,1 mg/ml Reactive Species,,Human,Rat,Mouse Host Species,,Rabbit Immunogen,,Synthesized peptide derived from human pr 1 * 50 µG
Catalog # GENOITN2676-50U-F
Supplier:  G-Biosciences
Target Protein,,P2RY6 Clonality Polyclonal Application,,WB,ELISA Recommended Dilution,,WB 1:500-2000 ELISA 1:5000-20000 Concentration,,1 mg/ml Reactive Species,,Human,Rat,Mouse Host Species,,Rabbit Immunogen,,Synthesized peptide derived from human pr 1 * 50 µG
Catalog # GENOITN2676-50U-F
Supplier:  G-Biosciences
Frequently Bought Together