Rodenticides Mixture 248 100 µg/mL in Acetonitrile 1 * 1 KIT
Catalog # EHERGS09000248AL
Supplier: EHRENSTORFER
Rodenticides Mixture 248 100 µg/mL in Acetonitrile 1 * 1 KIT
Catalog # EHERGS09000248AL
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Dispenser tips, PD-Tips II
Supplier: Brand
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Ibuprofen certified 1 * 0,25 g
Supplier: EHRENSTORFER