Deep well plate ,96- well , 2.2 ml , V- bottom , PP 1 * 50 items
Catalog # RATI6020221
Supplier: RATIOLAB
Deep well plate ,96- well , 2.2 ml , V- bottom , PP 1 * 50 items
Catalog # RATI6020221
Supplier: RATIOLAB
Frequently Bought Together
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Microtube racks
Supplier: RATIOLAB
Seals for ALPS™ 50 V heat sealer
Supplier: THERMO MATRIX TECHNOLOGIES
384-well clear polystyrene microplates, Corning®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Screw caps for T501 sample tubes
Supplier: Simport Scientific
Screw caps for SuperClear® microtubes
Supplier: Labcon
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Accessories for microplate modules
Supplier: Thermo Fisher Scientific
Multiwell cell culture plates, CELLSTAR®
Supplier: Greiner Bio-One
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES