10 reactions-The Thermo Scientific SureQuant AKT Pathway IP and MS Sample Preparation Module provides a complete reagent solution fo r the multiplexed enrichment and sample preparation of ten key AKT signaling pathway proteins for mass spectrometry ( 1 * 10 Assays
Catalog # PIERA40081
Supplier: Thermo Fisher Scientific
Frequently Bought Together