ContactUsLinkComponent
10 reactions-The Thermo Scientific SureQuant AKT Pathway Multiplex Panel (Relative Quantitation) provides a complete quantitation so lution for the enrichment and analysis of key AKT signaling pathway proteins using mass spectrometry (MS). 1 * 10 Assays
Catalog # PIERA40080
10 reactions-The Thermo Scientific SureQuant AKT Pathway Multiplex Panel (Relative Quantitation) provides a complete quantitation so lution for the enrichment and analysis of key AKT signaling pathway proteins using mass spectrometry (MS). 1 * 10 Assays
Catalog # PIERA40080
Frequently Bought Together