Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VENT-CAP wash bottle caps, PP, GL 25coloured blue, with short tube, PP 1 * 1 items
Catalog # VITL834121
Supplier:
VITLAB
Expand image
VENT-CAP wash bottle caps, PP, GL 25coloured blue, with short tube, PP 1 * 1 items
Catalog # VITL834121
Supplier:
VITLAB
Frequently Bought Together
Wash bottles, narrow neck, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Storage vials, round, Azlon®
Supplier: SciLabware
VENT-CAP wash bottle caps, PP,GL 25 coloured orange, with median 1 * 1 items
Supplier: VITLAB
VENT-CAP wash bottle caps, PP, GL 25coloured green, with median tube, PP 1 * 12 items
Supplier: VITLAB
Connection system GL 45
Supplier: SciLabware