ContactUsLinkComponent
Producer: Munktell Material: rayon ( viscose) Basis weight: 35g/ m Reel width: 490 mm Reel diam. : 240mm Reel length : 200m Ref 1.01306.0490200 1 * 1 items
Catalog # BINZ1.01306.049020
Supplier:  Ahlstrom-Munksjö
Producer: Munktell Material: rayon ( viscose) Basis weight: 35g/ m Reel width: 490 mm Reel diam. : 240mm Reel length : 200m Ref 1.01306.0490200 1 * 1 items
Catalog # BINZ1.01306.049020
Supplier:  Ahlstrom-Munksjö
Frequently Bought Together