ContactUsLinkComponent
Stopcock, Analytic, PTFE/25% glass fiber SJ18.8/38 For feeding of solvents and aerating the system. Less cross-contamination compared to standard stopcock For applications where grease should be avoided. Content: PTFE stopcock (no tubing included). 1 * 1 items
Catalog # BUCH11069607
Supplier:  BUCHI
Stopcock, Analytic, PTFE/25% glass fiber SJ18.8/38 For feeding of solvents and aerating the system. Less cross-contamination compared to standard stopcock For applications where grease should be avoided. Content: PTFE stopcock (no tubing included). 1 * 1 items
Catalog # BUCH11069607
Supplier:  BUCHI
Frequently Bought Together