ContactUsLinkComponent
Therm. for distillation range, similar to Kraemer-Spilker, similar to DIN 12785, enclosed scale, +60+210:0,2°C, length 480mm max., d iameter 7mm, cap. prismatic, colourless, red special liquid, immersion 90mm, richter-top finish, suitable for governm 1 * 1 items
Catalog # AMARD259500-GA
Supplier:  Amarell
Therm. for distillation range, similar to Kraemer-Spilker, similar to DIN 12785, enclosed scale, +60+210:0,2°C, length 480mm max., d iameter 7mm, cap. prismatic, colourless, red special liquid, immersion 90mm, richter-top finish, suitable for governm 1 * 1 items
Catalog # AMARD259500-GA
Supplier:  Amarell
Frequently Bought Together