ContactUsLinkComponent
Thermometer similar to BS 593, B160C/100, solid stem, white backed, 100+160:0,2°C, gallium filling, 400mm long, 100mm immersion, dur able pigment, with ice point scale 1 * 1 items
Catalog # AMARA403258-GA
Supplier:  Amarell
Thermometer similar to BS 593, B160C/100, solid stem, white backed, 100+160:0,2°C, gallium filling, 400mm long, 100mm immersion, dur able pigment, with ice point scale 1 * 1 items
Catalog # AMARA403258-GA
Supplier:  Amarell
Frequently Bought Together