ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 41 C, white backed, 98+152:0,2°C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 100mm, max.400x6-8mm, durable pigment, with Works Certificate with 1 test point 1 * 1 items
Catalog # AMARA300610-CB/CC4
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 41 C, white backed, 98+152:0,2°C, capillary tube specially coated inside, with non-wetting blue special liquid, immersion 100mm, max.400x6-8mm, durable pigment, with Works Certificate with 1 test point 1 * 1 items
Catalog # AMARA300610-CB/CC4
Supplier:  Amarell
Frequently Bought Together