ContactUsLinkComponent
Troemner OIML Precision Weights Weight Set(27), 2x2 kg-1 mg, OIML F1 1 * 1 items
Catalog # OHAU80780422
Supplier:  OHAUS
Troemner OIML Precision Weights Weight Set(27), 2x2 kg-1 mg, OIML F1 1 * 1 items
Catalog # OHAU80780422
Supplier:  OHAUS
Frequently Bought Together