Troemner OIML Precision Weights Weight Set(12), 500 mg-1 mg, OIML F1 1 * 1 items
OHAU80780408
: OHAUS
Troemner OIML Precision Weights Weight Set(12), 500 mg-1 mg, OIML F1 1 * 1 items
OHAU80780408
: OHAUS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK