ContactUsLinkComponent
Troemner OIML Precision Weights Weight Set(16), 5 kg-1 g, OIML E2, NVLAP 1 * 1 items
Catalog # OHAU80780400
Supplier:  OHAUS
Troemner OIML Precision Weights Weight Set(16), 5 kg-1 g, OIML E2, NVLAP 1 * 1 items
Catalog # OHAU80780400
Supplier:  OHAUS
Frequently Bought Together