ContactUsLinkComponent
Troemner OIML Precision Weights Wt Set(28), 5 kg-1 mg, OIML E2, NVLAP 1 * 1 items
Catalog # OHAU80780399
Supplier:  OHAUS
Troemner OIML Precision Weights Wt Set(28), 5 kg-1 mg, OIML E2, NVLAP 1 * 1 items
Catalog # OHAU80780399
Supplier:  OHAUS
Frequently Bought Together