ContactUsLinkComponent
Troemner OIML Precision Weights Weight Set (13), 1 kg-1 g, OIML E1 1 * 1 items
Catalog # OHAU30518000
Supplier:  OHAUS
Troemner OIML Precision Weights Weight Set (13), 1 kg-1 g, OIML E1 1 * 1 items
Catalog # OHAU30518000
Supplier:  OHAUS
Frequently Bought Together