ContactUsLinkComponent
BA81B MS w/100x Corded Binocular head, 45 inclined, 360 rotating Widefield eyepieces WF10X/18mm (paired) Reversed quadruple revolving nosepiece Achromatic objectives EA 4X, 10X, 40X S, 100X S Oil Coaxial coarse and fine focusing system with tension 1 * 1 items
Catalog # MOTI1100100450223
Supplier:  MOTIC
BA81B MS w/100x Corded Binocular head, 45 inclined, 360 rotating Widefield eyepieces WF10X/18mm (paired) Reversed quadruple revolving nosepiece Achromatic objectives EA 4X, 10X, 40X S, 100X S Oil Coaxial coarse and fine focusing system with tension 1 * 1 items
Catalog # MOTI1100100450223
Supplier:  MOTIC
Frequently Bought Together