ContactUsLinkComponent
Lactometer type Quevenne, 15-40:1/1, with therm. in the upper part, 0+40:1C, red or blue special liquid, ref. temp. 20C, reading above, 270mm long, government calibrated, with DKD-Certificate (DAkkS) with 3 test points 1 * 3 items
Catalog # AMARH841080-FL20C3
Dodavatel:  Amarell
Lactometer type Quevenne, 15-40:1/1, with therm. in the upper part, 0+40:1C, red or blue special liquid, ref. temp. 20C, reading above, 270mm long, government calibrated, with DKD-Certificate (DAkkS) with 3 test points 1 * 3 items
Catalog # AMARH841080-FL20C3
Dodavatel:  Amarell
Frequently Bought Together