Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 494.0 MPA.S (25.00C), RTM21 1 * 500 mL
Catalog # PARGRTM21
Supplier:
PARAGON SCIENTIFIC
Certificates
Expand image
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 494.0 MPA.S (25.00C), RTM21 1 * 500 mL
Catalog # PARGRTM21
Supplier:
PARAGON SCIENTIFIC
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, ready to use, 1 litre bottles, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Measuring beakers with handle
Supplier: BURKLE
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 1008 MPA.S (25.00C), RTM24 1 * 500 mL
Supplier: PARAGON SCIENTIFIC