Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 250.0 MPA.S (25.00C), RTM17 1 * 500 mL
Catalog # PARGRTM17
Supplier:
PARAGON SCIENTIFIC
Expand image
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 250.0 MPA.S (25.00C), RTM17 1 * 500 mL
Catalog # PARGRTM17
Supplier:
PARAGON SCIENTIFIC
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 75.19 MPA.S (25.00C), RTM13 1 * 500 mL
Supplier: PARAGON SCIENTIFIC