ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 6055 MPA.S (25.00C), RTM32 1 * 500 mL
Catalog # PARGRTM32
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 6055 MPA.S (25.00C), RTM32 1 * 500 mL
Catalog # PARGRTM32
Supplier:  PARAGON SCIENTIFIC
Frequently Bought Together