ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 0.9525 MPA.S (25.00C), RTM3 1 * 500 mL
Catalog # PARGRTM3
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 0.9525 MPA.S (25.00C), RTM3 1 * 500 mL
Catalog # PARGRTM3
Supplier:  PARAGON SCIENTIFIC