VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 30036 MPA.S (25.00C), RT30000 1 * 600 mL
PARGVIS-RT30K-600
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 30036 MPA.S (25.00C), RT30000 1 * 600 mL
PARGVIS-RT30K-600
Frequently Bought Together
Paracetamol
: EDQM
Nitenpyram Analytical standard, reference material, Fluka™
: Honeywell Chemicals
in Acetonitrile 1 * 1 mL
: EHRENSTORFER
VWR®, Test Sieves, 203×50 mm
: VWR Collection
Pirimicarb Analytical standard, reference material, Fluka™
: Honeywell Chemicals
Imazapyr certified 1 * 0,1 g
: EHRENSTORFER
Buffer Solutions
: Thermo Fisher Scientific
Cobalt standard solution, 1000 mg/L Co in 5% nitric acid (from Co), standard for AAS, Specpure®
: Thermo Fisher Scientific