ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 251.5 MPA.S (25.00C), RT250 1 * 600 mL
Catalog # PARGVIS-RT250-600
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 251.5 MPA.S (25.00C), RT250 1 * 600 mL
Catalog # PARGVIS-RT250-600
Supplier:  PARAGON SCIENTIFIC