Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 796.5 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN415
Supplier:
PARAGON SCIENTIFIC
Certificates
Expand image
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 796.5 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN415
Supplier:
PARAGON SCIENTIFIC
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Potassium dihydrogen phosphate 1/15 mol/l in aqueous solution, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
di-Ammonium oxalate solution Reag. Ph. Eur. 1005901
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
NISTmAb, Humanized IgG1k Monoclonal Antibody 1 * 1 items
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Supelco® PESTANAL® Organic Reference Standard, Piroctone olamine
Supplier: Merck
Conductivity standards, Potassium chloride, IUPAC
Supplier: Hach
VWR®, Density standards referenced at 25 °C
Supplier: VWR Chemicals
Ethanolamine, Supelco®
Supplier: Merck
Procymidone
Supplier: EHRENSTORFER
Pharmaceutical secondary standard, trimeprazine tartrate, (CRM), Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
2-FUCOSYLLACTOSE 1 * 200 mg
Supplier: USP