VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 30991 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN10200
Supplier: PARAGON SCIENTIFIC
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 30991 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN10200
Supplier: PARAGON SCIENTIFIC
Frequently Bought Together
Supelco® TraceCERT® Organic Reference Standard, Tebufenpyrad
Supplier: Merck
Elemental analysis standards, sulphur standards, VHG
Supplier: LGC Standards PROMOCHEM
BISOPROLOL FOR SYSTEM SUITABILITY 1 * 1 mg
Supplier: EDQM
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Nitrate standards for ISE calibration, Orion™
Supplier: Thermo Orion
Ofurace certified 1 * 0,1 g
Supplier: EHRENSTORFER
ACETYLTHIAMINE HYDROCHLORIDE, THIAMINE I 1 * 100 mg
Supplier: Merck
PROPYL GALLATE FOR ID AND ASSAY CRS 1 * 80 mg
Supplier: EDQM
Supelco® Organic Reference Standard, α-Arbutin
Supplier: Merck
Pesticide reference standard, Endrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
BREVETOXIN 3 SOLUTION 1 * 0,5 mL
Supplier: Merck
AVENANTHRAMIDE D, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
Pharmaceutical secondary standard, Guaiacol, (CRM), Supelco®
Supplier: Merck
Imazapyr certified 1 * 0,1 g
Supplier: EHRENSTORFER
Thiamphenicol certified 1 * 0,1 g
Supplier: EHRENSTORFER