ContactUsLinkComponent
Second display for Sartorius laboratory balances. To connect Sartorius lab balances of the product families Entris®, BSA, Practum®, Quintix® or Secura® via RS232 or USB interface. Scope of delivery: - Operation instruction, - Data cable USB-C to USB 1 * 1 items
Catalog # SARBYSD01
Supplier:  Sartorius Balances
Second display for Sartorius laboratory balances. To connect Sartorius lab balances of the product families Entris®, BSA, Practum®, Quintix® or Secura® via RS232 or USB interface. Scope of delivery: - Operation instruction, - Data cable USB-C to USB 1 * 1 items
Catalog # SARBYSD01
Supplier:  Sartorius Balances
Frequently Bought Together