ContactUsLinkComponent
GFP mRNA has been designed to produce high expression level of Green Fluorescent Protein. It is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm. 1 * 20 µG
Catalog # OZBIMRNA11-20
Supplier:  OZ BIOSCIENCES
GFP mRNA has been designed to produce high expression level of Green Fluorescent Protein. It is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm. 1 * 20 µG
Catalog # OZBIMRNA11-20
Supplier:  OZ BIOSCIENCES
Frequently Bought Together