ContactUsLinkComponent
Discharge valve for 25ml, 50ml or 100ml Varispenser 2(x) made of PFA, borosilicate glass, ceramic and platinum-iridium 1 * 1 items
Catalog # EPPE4966606.008
Supplier:  EPPENDORF
Discharge valve for 25ml, 50ml or 100ml Varispenser 2(x) made of PFA, borosilicate glass, ceramic and platinum-iridium 1 * 1 items
Catalog # EPPE4966606.008
Supplier:  EPPENDORF
Frequently Bought Together