ContactUsLinkComponent
Biological indicators for steam sterilization processes with G. stearothermophilus ATCC® 7953 spores inoculated on paper strips. 1 * 1 KIT
Catalog # LIOF91057
Supplier:  LIOFILCHEM
Biological indicators for steam sterilization processes with G. stearothermophilus ATCC® 7953 spores inoculated on paper strips. 1 * 1 KIT
Catalog # LIOF91057
Supplier:  LIOFILCHEM
Frequently Bought Together